Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium fluoride, Purified, ≥99%, Honeywell™

Click to view available options
Quantity:
2.5kg
7789-23-3
FK
58.097
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568
522689
CHEBI:66872
potassium;fluoride
[F-].[K+]
7789-23-3
FK
58.097
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride,potassiumfluoride,potassium fluoride kf,potassium fluorure,fluorure de potassium,potassium fluorure french,potassium fluoride on calcium fluoride,potassium fluoride, anhydrous,unii-9082wg1g3f,ccris 1568
522689
CHEBI:66872
potassium;fluoride
[F-].[K+]

Purified,≥99%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Linear Formula KF
UN Number 1812
Beilstein 3902818
Formula Weight 58.1g/mol
Percent Purity ≥99%
Grade Purified
Quantity 2.5kg
Chemical Name or Material Potassium Fluoride

P261-P280-P302 + P352 + P312-P304 + P340 + P312-P403 + P233

H301 + H311 + H331

EINECSNumber : 232-151-5