Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Fluoride Solution (50% w/w), Fisher Chemical

Click to view available options
Quantity:
20L
Packaging:
PolyPac™
13455-21-5,7732-18-5
FH4KO2
94.127
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate,unii-95r1d12heh,potassium dihydrate fluoride,potassium fluoride dihydrate, 98.5+%,potassium fluoride dihydrate, puratronic,potassium fluoride, dihydrate,potassium fluoride kf , dihydrate,acmc-1byyf,ksc497c4l,potassiumfluoridedihydrate
23708904
potassium;fluoride;dihydrate
O.O.[F-].[K+]
13455-21-5,7732-18-5
FH4KO2
94.127
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate,unii-95r1d12heh,potassium dihydrate fluoride,potassium fluoride dihydrate, 98.5+%,potassium fluoride dihydrate, puratronic,potassium fluoride, dihydrate,potassium fluoride kf , dihydrate,acmc-1byyf,ksc497c4l,potassiumfluoridedihydrate
23708904
potassium;fluoride;dihydrate
O.O.[F-].[K+]
Assay Percent Range 50% w/w
Concentration or Composition (by Analyte or Components) 50% w/w
Physical Form Liquid
Identification Pass Test
Packaging PolyPac™
Specific Gravity 1.28 to 1.33 (at 25°C)
Color Colorless
Quantity 20L
Chemical Name or Material Potassium Fluoride Solution, 50% (w/w)

DANGER!

Emergency Overview
Toxic by inhalation, in contact with skin and if swallowed. Causes severe respiratory tract irritation with possible burns. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1