Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Potassium fluorosulfate, 99.5%
Click to view available options
Quantity:
10g
50g
Specifications
Linear Formula | KSO3F |
UN Number | UN3260 |
Solubility Information | Hydrolysis. |
Formula Weight | 138.16 |
Physical Form | Powder/Lump |
Percent Purity | 99.5% |
Sensitivity | Moisture sensitive |
Color | White |
Melting Point | 350°C (decomposition) |
Quantity | 10g |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures