Potassium formate, 99% (water <2%), Alfa Aesar™

Manufacturer: Alfa Aesar A145510B

Quantity 1000g

 View more versions of this product

Catalog No. AAA145510B

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range 99%
    Molecular Formula HCO2K
    Formula Weight 84.12
    Chemical Name or Material Potassium Formate

    Specifications

    Formula Weight 84.12
    Quantity 1000g
    Melting Point 167°C
    TSCA TSCA
    MDL Number MFCD00013100
    Beilstein 3623272
    Merck Index 14,7633
    Solubility Information Soluble in water
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se
    RTECS Number LQ9625000
    Density 1.91
    Assay Percent Range 99%
    Molecular Formula HCO2K
    CAS 590-29-4
    Storage Note 1 Hygroscopic
    Chemical Name or Material Potassium Formate
    Assay 99%, (water <2%)

    Description

    Applications
    Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

    Solubility
    Soluble in water and alcohol. Insoluble in ether.

    Notes
    Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se

    Hygroscopic

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit