Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium formate, 99% (water less than 2%), Thermo Scientific Chemicals

Catalog Number AAA1455130
Click to view available options
Quantity:
250 g
1000 g
5000 g
590-29-4
CHKO2
84.115
MFCD00013100
WFIZEGIEIOHZCP-UHFFFAOYSA-M
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
2735122
CHEBI:63316
potassium;formate
C(=O)[O-].[K+]
590-29-4
CHKO2
84.115
MFCD00013100
WFIZEGIEIOHZCP-UHFFFAOYSA-M
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
2735122
CHEBI:63316
potassium;formate
C(=O)[O-].[K+]

Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 165°C to 168°C
Density 1.91
Assay Percent Range 99%
Quantity 250 g
Beilstein 3623272
Sensitivity Very Hygroscopic
Merck Index 14,7633
Solubility Information Soluble in water and alcohol; Insoluble in ether.
Formula Weight 84.12
Percent Purity 99%
Assay (water <2%)
Chemical Name or Material Potassium Formate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-677-9
RTECSNumber LQ9625000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit