Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium heptafluorotantalate(V), 99.7%

Manufacturer:  Alfa Aesar™ 4031609

Quantity 10g

 View more versions of this product

Catalog No. AA4031609


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16924-00-8
MFCD00042369
Tantalum potassium fluoride
392.14
UN3288
Potassium heptafluorotantalate(V)

Specifications

16924-00-8
392.14
UN3288
Decomposes in water; Slightly soluble in HF
10g
99.7%
Powder
MFCD00042369
Tantalum potassium fluoride
4.560g/mL
Potassium heptafluorotantalate(V)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit