Alfa Aesar™ Potassium hexabromoiridate(IV), 99.9% (metals basis), Ir 25.2% min

Manufacturer: Alfa Aesar™ 4101206

Quantity 5g

 View more versions of this product

Catalog No. AA4101206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19121-78-9
    Formula Weight 749.86
    MDL Number MFCD00054221
    Chemical Name or Material Potassium hexabromoiridate(IV)

    Specifications

    Formula Weight 749.86
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310-P301+P312-P406-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant/... container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    MDL Number MFCD00054221
    Health Hazard 1 Danger
    TSCA TSCA
    Color Black
    Quantity 5g
    CAS 19121-78-9
    Health Hazard 2 GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    Concentration Ir 25.2% min.
    Chemical Name or Material Potassium hexabromoiridate(IV)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H318-H290-H302
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.

    GHS P Statement
    P280-P305+P351+P338-P310-P301+P312-P406-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant/... container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit