Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexabromoiridate(IV), 99.9% (metals basis), Ir 25.2% min

Manufacturer:  Alfa Aesar™ 4101203

Quantity 1g

 View more versions of this product

Catalog No. AA4101203


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

19121-78-9
749.86
MFCD00054221
Potassium hexabromoiridate(IV)

Specifications

19121-78-9
749.86
Ir 25.2% min.
Hygroscopic
1g
99.9%, (metals basis)
Crystalline
MFCD00054221
Odorless
Black
Potassium hexabromoiridate(IV)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H290-H302
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P280-P305+P351+P338-P310-P301+P312-P406-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant/... container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit