Alfa Aesar™ Potassium hexachloroiridate(IV), Ir 39% min

Manufacturer: Alfa Aesar™ 1188803

Quantity 1g

 View more versions of this product

Catalog No. AA1188803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16920-56-2
    Formula Weight 483.12
    MDL Number MFCD00011369
    Chemical Name or Material Potassium hexachloroiridate(IV)

    Specifications

    Formula Weight 483.12
    MDL Number MFCD00011369
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline
    Sensitivity Hygroscopic
    CAS 16920-56-2
    Quantity 1g
    Concentration Ir 39% min.
    Chemical Name or Material Potassium hexachloroiridate(IV)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit