Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachloroosmate(IV), min 39% Os, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16871-60-6
Cl6K2Os
481.11
MFCD00011371
16871-60-6
Cl6K2Os
481.11
MFCD00011371
Linear Formula K2OsCl6
Merck Index 15, 7756
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
Formula Weight 481.11
Physical Form Powder
Percent Purity min 39% Os
Density 3.42
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
Color Bordeaux to Purple
Melting Point 600.0°C
Quantity 5g
Chemical Name or Material Potassium hexachloroosmate(IV), min 39% Os
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 240-893-6

TSCA : TSCA