Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexachloropalladate(IV), Pd 26.3% min

Manufacturer:  Alfa Aesar™ 1188414

Quantity 25g

 View more versions of this product

Catalog No. AA1188414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16919-73-6
MFCD00043081
Potassium hexachloropalladate(IV)
397.32
UN3077

Specifications

16919-73-6
397.32
UN3077
2.738g/mL
Hygroscopic
25g
Crystalline
MFCD00043081
Pd 26.3% min.
Odorless
Red
Potassium hexachloropalladate(IV)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : RT3353000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit