Alfa Aesar™ Potassium hexachloropalladate(IV), Pd 26.3% min

Manufacturer: Alfa Aesar™ 1188403

Quantity 1g

 View more versions of this product

Catalog No. AA1188403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16919-73-6
    Formula Weight 397.32
    MDL Number MFCD00043081
    UN Number UN3077
    Chemical Name or Material Potassium hexachloropalladate(IV)

    Specifications

    Formula Weight 397.32
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P310-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Quantity 1g
    Density 2.738g/mL
    Sensitivity Hygroscopic
    MDL Number MFCD00043081
    CAS 16919-73-6
    UN Number UN3077
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Physical Form Crystalline
    Color Red
    Concentration Pd 26.3% min.
    RTECS Number RT3353000
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302-H317-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Causes skin irritation.
    Chemical Name or Material Potassium hexachloropalladate(IV)

    GHS H Statement
    H318-H302-H317-H315
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Causes skin irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P310-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit