Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexachloroplatinate(IV), Pt 39.6%

Manufacturer:  Alfa Aesar™ 1216903

Quantity 1g

 View more versions of this product

Catalog No. AA1216903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16921-30-5
486.01
UN3288
Potassium hexachloroplatinate(IV)
K2PtCl6
MFCD00011389
14,7636

Specifications

16921-30-5
K2PtCl6
MFCD00011389
14,7636
Pt 39.6%
Odorless
Hygroscopic
250°C (dec.)
Potassium hexachloroplatinate(IV)
Crystalline Powder
486.01
UN3288
Soluble in hot water; Slightly soluble in cold water; Practically insoluble in alcohol
3.50g/mL
≥1.825
Yellow
1g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H334-H372-H318-H290-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : TP1650000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit