Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexachlororhenate(IV), 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA6211306
Click to view available options
Quantity:
1 g
5 g
16940-97-9
Cl6K2Re
477.10
MFCD00058746
HGCRXIXEXSEJGV-UHFFFAOYSA-H
potassium hexachlororhenate iv, dipotassium;hexachlororhenium 2-, dipotassium hexachlororheniumdiuide
10939720
dipotassium;hexachlororhenium(2-)
[K+].[K+].Cl[Re--](Cl)(Cl)(Cl)(Cl)Cl
16940-97-9
Cl6K2Re
477.10
MFCD00058746
HGCRXIXEXSEJGV-UHFFFAOYSA-H
potassium hexachlororhenate iv, dipotassium;hexachlororhenium 2-, dipotassium hexachlororheniumdiuide
10939720
dipotassium;hexachlororhenium(2-)
[K+].[K+].Cl[Re--](Cl)(Cl)(Cl)(Cl)Cl

Potassium hexachlororhenate is used in preparation of supported rhenium tricarbonyls which catalyze propylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Green
Physical Form Crystalline
Quantity 5 g
Linear Formula K2ReCl6
Solubility Information Soluble in water.
Formula Weight 477.12
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 3.34 g/mL
Assay (metals basis)
Chemical Name or Material Potassium hexachlororhenate (IV)
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-008-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit