Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium hexacyanocobaltate(III), 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Brown to Yellow |
Physical Form | Crystalline |
Quantity | 250 g |
Linear Formula | K3[Co(CN)6] |
Merck Index | 14,7637 |
Solubility Information | Soluble in water and acetic acid solutions; Insoluble in alcohol |
Formula Weight | 332.33 |
Percent Purity | ≥90% |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H311-H351-H302-H332-H317 Toxic in contact with skin. Suspected of causing cancer. Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P281-P308+P313-P501c |
EINECSNumber | 237-742-1 |
RTECSNumber | GF9470000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction