Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexacyanocobaltate(III), 95%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
13963-58-1
C6CoK3N6
332.34
MFCD00011391
13963-58-1
C6CoK3N6
332.34
MFCD00011391
Assay Percent Range 95%
Linear Formula K3[Co(CN)6]
Merck Index 15, 7758
Solubility Information Solubility in water: freely soluble. Other solubilities: insoluble in alcohol
Formula Weight 332.34
Physical Form Crystalline Solid
Percent Purity 95%
Density 1.8700g/mL
Packaging Glass bottle
Specific Gravity 1.87
Color Brown to Yellow
Quantity 500g
Chemical Name or Material Potassium hexacyanocobaltate(III), 95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-742-1

RTECSNumber : GF9470000

TSCA : TSCA