Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium hexacyanoferrate(II) trihydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
500g
1kg
6 x 500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | K4Fe(CN)6 · 3H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 422.39g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Quantity | 1kg |
Chemical Name or Material | Potassium Hexacyanoferrate(Ii) Trihydrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P273
H412
EINECSNumber : 237-722-2