Alfa Aesar™ Potassium hexacyanoplatinate(IV)

Manufacturer: Alfa Aesar™ 1267803

Quantity 1g

 View more versions of this product

Catalog No. AA1267803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16920-94-8
    Linear Formula K2Pt(CN)6
    Formula Weight 429.4
    MDL Number MFCD00058745
    UN Number UN1588
    Chemical Name or Material Potassium hexacyanoplatinate(IV)

    Specifications

    Formula Weight 429.4
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1588
    Odor Odorless
    MDL Number MFCD00058745
    Color White
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
    Physical Form Powder
    Linear Formula K2Pt(CN)6
    CAS 16920-94-8
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Potassium hexacyanoplatinate(IV)

    GHS H Statement
    H301-H311-H331-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit