Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min

Click to view available options
Quantity:
0.2g
2g
10g
339268-21-2
C6H2K4N6ORu
431.586
MFCD00168064
ZJQKAKIKDOERFU-UHFFFAOYSA-N
potassium hexacyanoruthenate ii hydrate,tetrapotassium hexacyanorutheniumtetrauide hydrate
71311369
tetrapotassium;ruthenium(2+);hexacyanide;hydrate
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.O.[K+].[K+].[K+].[K+].[Ru+2]
339268-21-2
C6H2K4N6ORu
431.586
MFCD00168064
ZJQKAKIKDOERFU-UHFFFAOYSA-N
potassium hexacyanoruthenate ii hydrate,tetrapotassium hexacyanorutheniumtetrauide hydrate
71311369
tetrapotassium;ruthenium(2+);hexacyanide;hydrate
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.O.[K+].[K+].[K+].[K+].[Ru+2]
Linear Formula K4Ru(CN)6·xH2O
UN Number UN1588
Solubility Information Soluble in water.
Formula Weight 413.57 (Anhydrous)
Physical Form Crystalline
Concentration Ru 23.0% min.
Color White
Quantity 0.2g
Chemical Name or Material Potassium hexacyanoruthenate(II) hydrate

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

EINECSNumber : 239-097-1

TSCA : No

Recommended Storage : Ambient temperatures