Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min

Manufacturer:  Alfa Aesar™ 1187604

Quantity 2g

 View more versions of this product

Catalog No. AA1187604


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

339268-21-2
413.57 (Anhydrous)
UN1588
K4Ru(CN)6·xH2O
MFCD00168064
Potassium hexacyanoruthenate(II) hydrate

Specifications

339268-21-2
K4Ru(CN)6·xH2O
MFCD00168064
Ru 23.0% min.
2g
Crystalline
413.57 (Anhydrous)
UN1588
White
Potassium hexacyanoruthenate(II) hydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit