Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexafluoroantimonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
16893-92-8
F6KSb
274.849
MFCD00043080
LQKKGPUEJBHVHZ-UHFFFAOYSA-H
potassium hexafluoroantimonate,acmc-20ajup,potassium ion hexafluorostibanuide,potassium hexafluoroantimonate min,potassium hexafluoroantimonate 1-,antimonate 1-,hexafluoro-, potassium 1:1 , oc-6-11
16688490
potassium;hexafluoroantimony(1-)
F[Sb-](F)(F)(F)(F)F.[K+]
16893-92-8
F6KSb
274.849
MFCD00043080
LQKKGPUEJBHVHZ-UHFFFAOYSA-H
potassium hexafluoroantimonate,acmc-20ajup,potassium ion hexafluorostibanuide,potassium hexafluoroantimonate min,potassium hexafluoroantimonate 1-,antimonate 1-,hexafluoro-, potassium 1:1 , oc-6-11
16688490
potassium;hexafluoroantimony(1-)
F[Sb-](F)(F)(F)(F)F.[K+]
Assay Percent Range 98%
Linear Formula KSbF6
Formula Weight 274.87
Physical Form Crystalline Powder
Elemental Analysis 97.5% min.
Packaging Glass bottle
Color White to Beige
Quantity 5g
Chemical Name or Material Potassium hexafluoroantimonate

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning