Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexafluoroarsenate(V), 99% (metals basis), Alfa aesar™

Manufacturer:  Alfa Aesar 1153206

Quantity 5g

 View more versions of this product

Catalog No. AA1153206


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

17029-22-0
99% (metals basis)
228.01
Potassium hexafluoroarsenate(V)
Powder
KAsF6
MFCD00042546

Specifications

17029-22-0
99% (metals basis)
White powder
MFCD00042546
White
Potassium hexafluoroarsenate(V)
Powder
KAsF6
228.01
Acrid
5g

Safety and Handling

SDS & Safety Documents

H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 241-102-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit