Alfa Aesar™ Potassium hexafluoronickelate(IV)

Manufacturer: Alfa Aesar™ 3943909

Quantity 10g

 View more versions of this product

Catalog No. AA3943909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17218-47-2
    Formula Weight 250.91
    MDL Number MFCD00083064
    UN Number UN3288
    Chemical Name or Material Potassium hexafluoronickelate(IV)

    Specifications

    Formula Weight 250.91
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3288
    MDL Number MFCD00083064
    CAS 17218-47-2
    Quantity 10g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H331-H350-H302-H312-H317
    Toxic if inhaled.
    May cause cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Chemical Name or Material Potassium hexafluoronickelate(IV)

    GHS H Statement
    H331-H350-H302-H312-H317
    Toxic if inhaled.
    May cause cancer.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit