Learn More
Alfa Aesar™ Potassium hexafluoronickelate(IV)
Manufacturer:  Alfa Aesar™ 3943904
Quantity 2g
 View more versions of this product
Catalog No. AA3943904
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
17218-47-2
MFCD00083064
Potassium hexafluoronickelate(IV)
250.91
UN3288
Specifications
17218-47-2
250.91
UN3288
Potassium hexafluoronickelate(IV)
Powder
MFCD00083064
2g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H331-H350-H302-H312-H317
Toxic if inhaled.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.