Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium hexafluorophosphate, 98%

Manufacturer:  Alfa Aesar™ A1208018

Quantity 50g

 View more versions of this product

Catalog No. AAA1208018


  Check Availability
Add to cart

Specifications

Description

Applications
Potassium hexafluorophosphate is used to prepare 1-butyl-3-methylimidazolium hexaflurorphospahte and other organic hexafluorophospahte compounds such as alkylaryliodonium hexafluorophopahtes and triarylsulfonium hexafluorophosphates. It is also used in the cationic polymerization reactions as photo initiators. It involves in producing catalyst for peptide coupling reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and oxidizing agents.
Description & Specifications

Chemical Identifiers

17084-13-8
184.07
UN3260
KPF6
MFCD00011412
Potassium hexafluorophosphate

Specifications

17084-13-8
184.07
UN3260
575°C
50g
98%
KPF6
MFCD00011412
Odorless
3.0 to 7.0
Potassium hexafluorophosphate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit