Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexafluorosilicate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 89023A7

Quantity 5kg

 View more versions of this product

Catalog No. AA89023A7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

16871-90-2
99%
220.28
Potassium fluorosilicate
Powder
K2SiF6
MFCD00040666
Potassium hexafluorosilicate

Specifications

16871-90-2
99%
220.28
Potassium fluorosilicate
5kg
Powder
K2SiF6
MFCD00040666
White
Potassium hexafluorosilicate

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 240-896-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit