Learn More
Potassium hydrogen carbonate, ACS, 99.7-100.5% (dried basis), Thermo Scientific Chemicals
Description
Potassium hydrogen carbonate is used in foods to regulate pH, as a fire suppression agent (BC dry chemical) and as a buffering agent in medications. It acts as an additive in winemaking and is an active component in club soda. Furthermore, it finds use in crops for the neutralization of acidic soils. It is also used as a source of carbon dioxide for leavening in baking. It is utilized to inhibit the growth of aspergillus parasiticus in czapek's agar. It is employed as a catalyst in synthetic fiber polymerization and olefin dehydrogenation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Granules |
Quantity | 100 g |
Assay Percent Range | 99.7-100.5% |
Solubility Information | Slightly soluble in water. Practically insoluble in alcoholSoluble in water. Insoluble in alcohol. |
IUPAC Name | potassium hydrogen carbonate |
Formula Weight | 100.12 |
Grade | ACS |
Odor | Odorless |
Appearance | White crystalline granules |
Show More |
Safety and Handling
EINECSNumber | 206-059-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.