Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydrogen carbonate, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
298-14-6
CHKO3
100.12
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium bicarbonate,potassium hydrogen carbonate,potassium hydrogencarbonate,potassium acid carbonate,monopotassium carbonate,carbonic acid, monopotassium salt,unii-hm5z15lebn,potassiumbicarbonate,ccris 3510,khco3
516893
CHEBI:81862
potassium;hydrogen carbonate
C(=O)(O)[O-].[K+]
298-14-6
CHKO3
100.12
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium bicarbonate,potassium hydrogen carbonate,potassium hydrogencarbonate,potassium acid carbonate,monopotassium carbonate,carbonic acid, monopotassium salt,unii-hm5z15lebn,potassiumbicarbonate,ccris 3510,khco3
516893
CHEBI:81862
potassium;hydrogen carbonate
C(=O)(O)[O-].[K+]
Linear Formula KHCO3
Ammonium 5ppm max
Merck Index 15,773
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C),practocally insoluble in alcohol,soluble in potassium carbonate solution
Formula Weight 100.12
Grade ACS Reagent
Chloride 0.001% max
Insoluble Matter 0.01% max
Packaging Plastic bottle
Phosphate 5ppm max
Solubility Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution
Melting Point 292.0°C
Quantity 100g
Chemical Name or Material Potassium hydrogen carbonate
Show More Show Less

EINECSNumber : 206-059-