Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium hydrogen diiodate, 99.85%, extra pure, ACROS Organics™
Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
Description
Oxidation reagent
Specifications
Additional Information | chloride, chlorate, bromide, bromate (as Cl): < 0.02% |
Assay Percent Range | 99.8% min. (Iodometry) |
Linear Formula | KH(IO3)2 |
Solubility Information | Solubility in water: 13g/L (15°C) |
Formula Weight | 389.9 |
Physical Form | Crystals or Powder |
Percent Purity | 99.85% |
Grade | Extra Pure |
Chloride | 0.02% max |
Insoluble Matter | 0.01% max |
Show More |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Ri
GHS Signal Word: Danger
EINECSNumber : 236-650-9
TSCA : TSCA