Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS, 99.95-105%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
877-24-7
C8H5KO4
204.22
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium 2-carboxybenzoate
[K+].OC(=O)C1=CC=CC=C1C([O-])=O
877-24-7
C8H5KO4
204.22
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium 2-carboxybenzoate
[K+].OC(=O)C1=CC=CC=C1C([O-])=O

P1190, 877-24-7, C8H5O4K

Spectrum™ Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS is used in analytical chemistry as a disintegrating agent can be used as a buffering agent (in combination with hydrochloric acid (HCl) or sodium hydroxide (NaOH). As an ACS grade Reagent, Spectrum Chemical manufactured Potassium Hydrogen Sulfate is used as the quality standard against which other Potassium Hydrogen Sulfate substances are graded. it has met the toughest regulatory standards for quality and pureness.

Packaging Poly Bottle
Quantity 25 g
pH 4.00 to 4.02
CAS Max % 1
Percent Purity 99.95 to 100.05%
Grade ACS