Learn More
Potassium hydrogen phthalate, primary standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Description
As a primary standard for preparing volumetric alkali solutions and as a buffer in pH determinations. Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters. It acts as a buffer in pH determination. It finds application as a useful standard for total organic carbon testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 297°C (decomposition) |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 99.95-100.05% |
Beilstein | 3637128 |
Solubility Information | Soluble in water. Slightly soluble in alcohol. |
Formula Weight | 204.23 |
Grade | ACS Reagent |
Density | 1.636 g/mL |
Show More |
Safety and Handling
EINECSNumber | 212-889-4 |
RTECSNumber | CZ4326000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.