Alfa Aesar™ Potassium hydrotris(1-pyrazolyl)borate

Manufacturer: Alfa Aesar™ 3943803

Quantity 1g

 View more versions of this product

Catalog No. AA3943803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18583-60-3
    Formula Weight 252.13
    MDL Number MFCD00040346
    Chemical Name or Material Potassium hydrotris(1-pyrazolyl)borate

    Specifications

    Formula Weight 252.13
    MDL Number MFCD00040346
    Quantity 1g
    Physical Form Crystalline
    CAS 18583-60-3
    Melting Point 185° to 189°C
    Chemical Name or Material Potassium hydrotris(1-pyrazolyl)borate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit