Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.005 Normal in Methanol, Reagents

Catalog No. p-200021610
Click to view available options
Quantity:
1 L
4 L

Potassium Hydroxide, 0.005 Normal in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1, 1310-58-3, 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name methanol
SMILES CO

Specifications

pH >12
Color Colorless
Boiling Point 65°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.005 Normal in Methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.