Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.010 Normal, Reagents

Catalog No. 01334085
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
01-334-085 4 L
01-334-086 1 L
2 options

Catalog No. 01-334-085

Supplier: Reagents Holdings Llc CP6051004A

Only null left
Add to Cart
Add to Cart

Potassium Hydroxide, 0.010 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name water
SMILES O

Specifications

pH 12
Color Colorless
Boiling Point 100°C
Quantity 4 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0006
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.010 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.