Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol, Reagents

Catalog No. 01334236
Change view
Click to view available options
Quantity:
500 mL
4 L
Catalog No. Quantity
01-334-236 500 mL
01-334-237 4 L
2 options

Catalog No. 01-334-236

Supplier: Reagents Holdings Llc CP199300500A

Only null left
Add to Cart
Add to Cart

Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1 , 1310-58-3 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name methanol
SMILES CO

Specifications

pH 12
Color Colorless
Boiling Point 65°C
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.0100 Normal (N/100) in Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.