Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.1N in Methanol Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle

Chemical Identifiers

1310-58-3
HKO
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
CHEBI:32035

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Flash Point 12.00°C
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Formula Weight 56.11
Packaging Cubetainer
Specific Gravity 1L = 0.80kg
Chemical Name or Material Potassium Hydroxide, 0.1N in Methanol Volumetric Solution
Grade BAKER ANALYZED™ Reagent