Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, 0.1N in Methanol Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
1310-58-3
HKO
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

High quality chemicals for laboratory and specialized industrial use

Quantity 1L
Flash Point 12.00°C
Formula Weight 56.11
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 0.80kg
Chemical Name or Material Potassium Hydroxide, 0.1N in Methanol Volumetric Solution