Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium hydroxide, 0.1N Standardized Solution, Alfa Aesar™
Click to view available options
Quantity:
1L
4L
20L
Specifications
Appearance | Colorless liquid |
Solubility Information | Freely soluble in water. Soluble in ethanol. |
Formula Weight | 56.11 |
Physical Form | Liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Color | Colorless |
Quantity | 1L |
Chemical Name or Material | Potassium hydroxide |
Safety and Handling
H315-H319: Causes skin irritation. Causes serious eye irritation.
EINECSNumber : 215-181-3
TSCA : Yes
Recommended Storage : Ambient temperatures