Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Color | Colorless |
Assay Percent Range | 0.2000 ± 0.0005 N at 20°C |
Solubility Information | Solubility in Water: Infinite |
Quantity | 500 mL |
Odor | Characteristic alcohol odor |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Nonaqueous Solution |
Physical Form | Liquid |
Chemical Name or Material | Potassium Hydroxide, 0.2N in Isopropyl Alcohol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction