Alfa Aesar™ Potassium hydroxide, 1.0N Standardized Solution

Manufacturer: Alfa Aesar™ 35592M4

Quantity 20L

 View more versions of this product

Catalog No. AA35592M4

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1310-58-3
    Formula Weight 56.11
    MDL Number MFCD00003553
    UN Number UN1814
    Chemical Name or Material Potassium hydroxide, Standardized Solution

    Specifications

    Formula Weight 56.11
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    Color Colorless
    Physical Form Liquid
    Concentration 1.0N
    MDL Number MFCD00003553
    UN Number UN1814
    Health Hazard 1 Danger
    TSCA TSCA
    Quantity 20L
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: POTASSIUM HYDROXIDE, SOLUTION
    CAS 1310-58-3
    Chemical Name or Material Potassium hydroxide, Standardized Solution

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit