Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, ACS Reagent, pellets, ≥85%, Honeywell Fluka™

Click to view available options
Quantity:
25g
500g
1kg
2.5kg
3.5kg
6 x 500g
12kg
1310-58-3
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

ACS Reagent, pellets,≥85%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula KOH
UN Number 1813
Formula Weight 56.11g/mol
Percent Purity ≥85%
Grade ACS Reagent
Vapor Pressure 1mmHg (719°C)
Quantity 12kg
Chemical Name or Material Potassium Hydroxide

P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H302-H314

EINECSNumber : 215-181-3

RTECSNumber : TT2100000