Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, BiotechGrade, 86%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]

P1317, 1310-58-3, KOH

Spectrum™ Potassium Hydroxide, Pellets, BiotechGrade is an inorganic compound commonly referred to as Caustic Potash. A colorless solid chemical compound and is a strong base. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Grade Biotech
CAS Max % 1
Appearance Pure, white pellet
Percent Purity 86%
Heavy Metals 3e-005
Iron (Fe) 3e-006
Lead (Pb) 1e-005
Mercury (Hg) 1e-007
Odor Odorless
Packaging Poly Bottle
Quantity 2.5 kg
Show More Show Less