Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, FCC, 85-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]

P1316, 1310-58-3, KOH

Spectrum™ Potassium Hydroxide, Pellets, FCC is used as a rinse or chemical peel for fruits and vegetables, a stabilizer to prolong shelf-life, and as a thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Percent Purity 85 to 100.5%
Lead (Pb) 2 mg/kg
Mercury (Hg) 0.1 mg/kg
Packaging Poly Bottle
Quantity 4 x 2.5 kg