Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide, Pellets, NF, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
[OH-].[K+]

PO180, 1310-58-3, KOH

Spectrum™ Potassium Hydroxide, Pellets, NF is used as a pH modifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade NF
CAS Max % 1
Packaging Poly Pail
Sodium (Na) 0.01
Quantity 12 kg