Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, pure, 8N solution in water, ACROS Organics™

Click to view available options
Quantity:
500mL
2L
10L
Packaging:
Plastic bottle
Plastic drum
1310-58-3,7732-18-5
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3,7732-18-5
HKO
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
Linear Formula KOH
Merck Index 15,7761
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
Formula Weight 56.11
Physical Form Liquid
Grade Pure
Composition 448g KOH / l H2O
Concentration 7.992 to 8.008N (20°C)
Density 1.3260g/mL
Packaging Plastic drum
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
Specific Gravity 1.326
Color Colorless
Quantity 10L
Chemical Name or Material Potassium hydroxide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-181-3