Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Honeywell Fluka™

Click to view available options
Quantity:
500mL
1L
2.5L
6 x 500mL
4 x 2.5L
6 x 1L

Volumetric, 0.5 M KOH in ethanol (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

CAS 1310-58-3
Concentration 0.5M in ethanol
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
UN Number UN2924
Synonym potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.105
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Flash Point 11°C (51.8°F)
Quantity 500mL
Chemical Name or Material Potassium hydroxide solution
Show More Show Less

P210-P260-P280-P305 + P351 + P338 + P310-P370 + P378-P403 + P235