Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Honeywell Fluka™

Click to view available options
Quantity:
500mL
1L
2.5L
4 x 2.5L
6 x 1L

Volumetric, 0.5 M KOH in ethanol (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

CAS 1310-58-3
Concentration 0.5M in ethanol
Molecular Formula HKO
Linear Formula KOH
MDL Number MFCD00003553
UN Number UN2924
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.105
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Flash Point 11°C (51.8°F)
Quantity 500mL
Chemical Name or Material Potassium hydroxide solution
Show More Show Less

P210-P260-P280-P305 + P351 + P338 + P310-P370 + P378-P403 + P235