Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution, 45%(w/w), BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
19L
Packaging:
Jerrican (Poly Pail)
Poly Bottle
1310-58-3
HKO
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.105
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Quantity 19L
Formula Weight 56.11
Percent Purity 45% (w/w)
Grade BAKER ANALYZED™ Reagent
Packaging Jerrican (Poly Pail)
Specific Gravity 1L = 1.45kg
Chemical Name or Material Potassium Hydroxide Solution