Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Hydroxide Solution (CO2 Absorbent/Certified), Fisher Chemical

Click to view available options
Quantity:
1L
Packaging:
Poly Bottle
1310-58-3,7732-18-5
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3,7732-18-5
HKO
56.105
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide,caustic potash,potash lye,potassium hydrate,hydroxyde de potassium,potassa,potasse caustique,potassium hydroxide k oh,potassium hydroxide solution,caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

For determination of carbon dioxide in gas analysis

Quantity 1L
Assay Percent Range 36 to 42 %
Identification Pass Test
Formula Weight 56.11
Physical Form Liquid
Percent Purity 36 to 42%
Grade Certified
Packaging Poly Bottle
Suitability Pass Test
Chemical Name or Material Potassium Hydroxide Solution, CO2 Absorbent