Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodate, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
7758-05-6, 7732-18-5
IKO3
214
MFCD00011406
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate
[K+].[O-][I](=O)=O
7758-05-6, 7732-18-5
IKO3
214
MFCD00011406
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate
[K+].[O-][I](=O)=O
Oxidation reagent
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula KIO3
Merck Index 15, 7763
Arsenic (As) 3ppm max.
Solubility Information Solubility in water: soluble
Formula Weight 214
Physical Form Crystalline Powder
Percent Purity 99.50%
Grade Analysis
Chlorate 100ppm max.
Density 3.9300g/mL
Heavy Metals (as Pb) 10ppm max.
Iodine (I) 20ppm max.
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 3.93
Color White
Melting Point 560°C
Quantity 500g
Chemical Name or Material Potassium iodate, p.a.
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-831-9

RTECSNumber : NN1350000

TSCA : TSCA